missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-pentyn-3-ol 98.0+%, TCI America™

Catalog No. M039625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M039625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M039625ML Supplier TCI America Supplier No. M039625ML
Only null left

Chemical Identifiers

CAS 77-75-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00004479
InChI Key QXLPXWSKPNOQLE-UHFFFAOYNA-N
Synonym 3-methyl-1-pentyn-3-ol, meparfynol, methylpentynol, methylparafynol, seral, atemorin, melpintol, 1-pentyn-3-ol, 3-methyl, allotropal, methylpentinol
PubChem CID 6494
IUPAC Name 3-methylpent-1-yn-3-ol
SMILES CCC(C)(O)C#C

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 mL
UN Number 1987
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-pentyn-3-ol
EINECSNumber (2)-0264
RTECSNumber SC4900000
TSCA Yes