accessibility menu, dialog, popup

UserName

CH3CH3OHHC

CAS RN 77-75-8

IUPAC Name: 3-methylpent-1-yn-3-ol
Synonyms: methylpentynol ethinylmethylethylcarbinol 3-methyl-pentin-(1)-ol-(3) N-oblivon 3-metil-pentin-3-ol olfine P methylparafynol (substance) scherer brand of methylpentynol 3-methylpentin-3-ol anti-stress
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: QXLPXWSKPNOQLE-UHFFFAOYNA-N
SMILES: CCC(C)(O)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results