missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-pentene 98.0+%, TCI America™

$67.11 - $67.11

Chemical Identifiers

CAS 760-20-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009339
InChI Key LDTAOIUHUHHCMU-UHFFFAOYSA-N
Synonym 3-methyl-1-pentene, 1-pentene, 3-methyl, ethene, trimer, 2-vinylbutane, sec-butylethene, 3-methylpentene-1, pentene, 3-methyl, 1-pentene,3-methyl, acmc-1bmfr
PubChem CID 12969
IUPAC Name 3-methylpent-1-ene
SMILES CCC(C)C=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M05751G
View Documents
TCI America
M05751G
1 g
Each for $67.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

760-20-3
84.162
LDTAOIUHUHHCMU-UHFFFAOYSA-N
12969
CCC(C)C=C
C6H12
MFCD00009339
3-methyl-1-pentene, 1-pentene, 3-methyl, ethene, trimer, 2-vinylbutane, sec-butylethene, 3-methylpentene-1, pentene, 3-methyl, 1-pentene,3-methyl, acmc-1bmfr
3-methylpent-1-ene

Specifications

760-20-3
54°C
MFCD00009339
2288
LDTAOIUHUHHCMU-UHFFFAOYSA-N
3-methylpent-1-ene
12969
≥98.0% (GC)
3-Methyl-1-pentene
Colorless
C6H12
1 g
3-methyl-1-pentene, 1-pentene, 3-methyl, ethene, trimer, 2-vinylbutane, sec-butylethene, 3-methylpentene-1, pentene, 3-methyl, 1-pentene,3-methyl, acmc-1bmfr
CCC(C)C=C
84.162
84.16
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0031

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'