accessibility menu, dialog, popup

UserName

CH3H3CH2C

CAS RN 760-20-3

IUPAC Name: 3-methylpent-1-ene
Synonyms: 3-methyl-1-pentene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: LDTAOIUHUHHCMU-UHFFFAOYNA-N
SMILES: CCC(C)C=C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)