missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-pentanol 99.0+%, TCI America™

Catalog No. p-7132259
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M060010ML 10 mL
M0600100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M060010ML Supplier TCI America Supplier No. M060010ML

Chemical Identifiers

CAS 589-35-5
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002937
InChI Key IWTBVKIGCDZRPL-UHFFFAOYSA-N
Synonym 3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol
PubChem CID 11508
ChEBI CHEBI:87381
IUPAC Name 3-methylpentan-1-ol
SMILES CCC(C)CCO

Specifications

Color Yellow
Boiling Point 152°C
Quantity 10 mL
UN Number 1987
Formula Weight 102.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-pentanol
EINECSNumber (2)-0217
TSCA Yes