accessibility menu, dialog, popup

UserName

H3CCH3OH

CAS RN 589-35-5

IUPAC Name: 3-methylpentan-1-ol
Synonyms: 3-methyl-1-pentanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: IWTBVKIGCDZRPL-UHFFFAOYNA-N
SMILES: CCC(C)CCO
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)