missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-cyclohexene 93.0+%, TCI America™

Catalog No. M103425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M103425ML 25 mL
M1034500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M103425ML Supplier TCI America Supplier No. M103425ML

Chemical Identifiers

CAS 591-48-0
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00001571
InChI Key UZPWKTCMUADILM-UHFFFAOYSA-N
Synonym 3-methyl-1-cyclohexene, cyclohexene, 3-methyl, 3-methyl cyclohexene, 3-methyl-cyclohexene, 3-methylcyclohexene-1, 3-methylcyclohex-1-ene, acmc-1atr1
PubChem CID 11573
IUPAC Name 3-methylcyclohexene
SMILES CC1CCCC=C1

Specifications

Color Colorless
Boiling Point 105°C
Quantity 25 mL
UN Number 3295
Formula Weight 96.17
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-cyclohexene