accessibility menu, dialog, popup

UserName

CAS RN 591-48-0

IUPAC Name: 3-methylcyclohex-1-ene
Synonyms: 3-methyl-1-cyclohexene
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: UZPWKTCMUADILM-UHFFFAOYNA-N
SMILES: CC1CCCC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)