Learn More
3-Methyl-1-butanol, ACS, 98.5+%
CAS: 123-51-3 | C5H12O | 88.15 g/mol
Supplier: Thermo Scientific Chemicals 036716K2
Description
3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Methyl-1-butanol | |
| -117°C | |
| 130°C to 132°C | |
| 98.5+% | |
| C5H12O | |
| MFCD00002934 | |
| UN1105 | |
| 14,5195 | |
| Miscible with water,alcohol,ether,benzene,chloroform,petroleum ether,glacial acetic acid,acetone and oils. | |
| CC(C)CCO | |
| 88.15 | |
| CHEBI:15837 | |
| ≥98.5% | |
| Liquid |
| 123-51-3 | |
| 0.809 g/mL | |
| 45°C (113°F) | |
| Unpleasant | |
| 1.406 | |
| 1 L | |
| 1718835 | |
| 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
| PHTQWCKDNZKARW-UHFFFAOYSA-N | |
| 3-methylbutan-1-ol | |
| 31260 | |
| 88.15 | |
| ACS |
Chemical Identifiers
| 123-51-3 | |
| 88.15 | |
| PHTQWCKDNZKARW-UHFFFAOYSA-N | |
| 31260 | |
| 3-methylbutan-1-ol |
| C5H12O | |
| MFCD00002934 | |
| 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
| CHEBI:15837 | |
| CC(C)CCO |
Safety and Handling
GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H332-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 204-633-5
RTECSNumber : EL5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only