Learn More
3-Methyl-1-butanol, ACS, 98.5+%
Description
3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 117°C |
| Density | 0.809 |
| Boiling Point | 130°C to 132°C |
| Flash Point | 45°C (113°F) |
| Assay Percent Range | 98.5+% |
| Refractive Index | 1.406 |
| Quantity | 500 mL |
| UN Number | UN1105 |
| Beilstein | 1718835 |
| Merck Index | 14,5195 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H226-H332-H335 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 204-633-5 |
| RTECSNumber | EL5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only