missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1,5-pentanediol 97.0+%, TCI America™
$63.38 - $197.59
Chemical Identifiers
| CAS | 4457-71-0 |
|---|---|
| Molecular Formula | C6H14O2 |
| Molecular Weight (g/mol) | 118.176 |
| MDL Number | MFCD00039547 |
| InChI Key | SXFJDZNJHVPHPH-UHFFFAOYSA-N |
| PubChem CID | 20524 |
| IUPAC Name | 3-methylpentane-1,5-diol |
| SMILES | CC(CCO)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M071025ML
|
TCI America
M071025ML |
25 mL |
Each for $63.38
|
|
|||||
|
M0710500ML
|
TCI America
M0710500ML |
500 mL |
Each for $197.59
|
|
|||||
Chemical Identifiers
| 4457-71-0 | |
| 118.176 | |
| SXFJDZNJHVPHPH-UHFFFAOYSA-N | |
| 3-methylpentane-1,5-diol |
| C6H14O2 | |
| MFCD00039547 | |
| 20524 | |
| CC(CCO)CCO |
Specifications
| 4457-71-0 | |
| 150°C | |
| MFCD00039547 | |
| 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference | |
| CC(CCO)CCO | |
| 118.176 | |
| 118.18 | |
| Liquid |
| Colorless | |
| C6H14O2 | |
| 25 mL | |
| SXFJDZNJHVPHPH-UHFFFAOYSA-N | |
| 3-methylpentane-1,5-diol | |
| 20524 | |
| ≥97.0% (GC) | |
| 3-Methyl-1,5-pentanediol |
Safety and Handling
EINECSNumber : (2)-0240
RTECSNumber : SA0800000
TSCA : Yes