missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1,5-pentanediol, 99%
CAS: 4457-71-0 | C6H14O2 | 118.176 g/mol
$129.22 - $488.99
Chemical Identifiers
| CAS | 4457-71-0 |
|---|---|
| Molecular Formula | C6H14O2 |
| Molecular Weight (g/mol) | 118.176 |
| MDL Number | MFCD00039547 |
| InChI Key | SXFJDZNJHVPHPH-UHFFFAOYSA-N |
| Synonym | 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference |
| PubChem CID | 20524 |
| IUPAC Name | 3-methylpentane-1,5-diol |
| SMILES | CC(CCO)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2054606
|
Thermo Scientific Chemicals
B2054606 |
5 g |
Each for $129.22
|
|
|||||
|
AAB2054614
|
Thermo Scientific Chemicals
B2054614 |
25 g |
Each for $488.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4457-71-0 | |
| 118.176 | |
| SXFJDZNJHVPHPH-UHFFFAOYSA-N | |
| 20524 | |
| CC(CCO)CCO |
| C6H14O2 | |
| MFCD00039547 | |
| 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference | |
| 3-methylpentane-1,5-diol |
Specifications
| 4457-71-0 | |
| 143°C (289°F) | |
| 1.454 | |
| 5 g | |
| 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference | |
| CC(CCO)CCO | |
| 118.176 | |
| 118.18 | |
| 3-Methyl-1,5-pentanediol |
| 0.97 | |
| C6H14O2 | |
| MFCD00039547 | |
| 1697331 | |
| SXFJDZNJHVPHPH-UHFFFAOYSA-N | |
| 3-methylpentane-1,5-diol | |
| 20524 | |
| 99% |
Safety and Handling
EINECSNumber : 224-709-1
RTECSNumber : SA0800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only