missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-(3'-sulfoamidophenyl)-5-pyrazolone 95.0+%, TCI America™

Catalog No. M065925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M065925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M065925G Supplier TCI America Supplier No. M065925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-29-2
Molecular Formula C10H11N3O3S
Molecular Weight (g/mol) 253.28
MDL Number MFCD00035707
InChI Key ASVVGQURNHNITH-UHFFFAOYSA-N
PubChem CID 66634
IUPAC Name 3-(3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzene-1-sulfonamide
SMILES CC1=NN(C(=O)C1)C1=CC=CC(=C1)S(N)(=O)=O

Specifications

Melting Point 196°C
Quantity 25 g
Formula Weight 253.28
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-1-(3′-sulfoamidophenyl)-5-pyrazolone
EINECSNumber (5)-0358
TSCA Yes