missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,3-butanediol 98.0+%, TCI America™

Catalog No. M081825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M081825ML 25 mL
M0818500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M081825ML Supplier TCI America Supplier No. M081825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2568-33-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00059655
InChI Key XPFCZYUVICHKDS-UHFFFAOYSA-N
Synonym 3-methyl-1,3-butanediol, 1,3-butanediol, 3-methyl, isopentyldiol, 2-methyl-2,4-butanediol, unii-19nol5474q, 2,4-dihydroxy-2-methylbutane, 3-methyl-butane-1,3-diol, 1,3-butanediol,3-methyl, 3-methyl-1,3-butandiol, 3-methylbutan-1,3-diol
PubChem CID 247470
IUPAC Name 3-methylbutane-1,3-diol
SMILES CC(C)(CCO)O

Specifications

Color Colorless
Boiling Point 203°C
Quantity 25 mL
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1,3-butanediol
EINECSNumber (2)-0240
TSCA Yes