accessibility menu, dialog, popup

UserName

CAS RN 2568-33-4

CH3CH3OHHO

CAS RN 2568-33-4

IUPAC Name: 3-methylbutane-1,3-diol
Synonyms: 2,4-dihydroxy-2-methylbutane
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: XPFCZYUVICHKDS-UHFFFAOYSA-N
SMILES: CC(C)(O)CCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results