missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1,2-dihydropyridin-2-one, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 1003-56-1 |
|---|---|
| Molecular Formula | C6H7NO |
| Molecular Weight (g/mol) | 109.128 |
| InChI Key | MVKDNXIKAWKCCS-UHFFFAOYSA-N |
| Synonym | 3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone |
| PubChem CID | 70481 |
| IUPAC Name | 3-methyl-1H-pyridin-2-one |
| SMILES | CC1=CC=CNC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
CD09405DA
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 1003-56-1 | |
| 109.128 | |
| 3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone | |
| 3-methyl-1H-pyridin-2-one |
| C6H7NO | |
| MVKDNXIKAWKCCS-UHFFFAOYSA-N | |
| 70481 | |
| CC1=CC=CNC1=O |
Specifications
| 1003-56-1 | |
| C6H7NO | |
| MVKDNXIKAWKCCS-UHFFFAOYSA-N | |
| 3-methyl-1H-pyridin-2-one | |
| 70481 | |
| 97% |
| 97% | |
| 3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone | |
| CC1=CC=CNC1=O | |
| 109.128 | |
| 109.13 | |
| 3-Methyl-1, 2-dihydropyridin-2-one |