missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophenol, 97%

CAS: 15570-12-4 | C7H8OS | 140.2 g/mol

Supplier:  Thermo Scientific Chemicals A1064806

 View more versions of this product

Catalog No. AAA1064806


Only null left
Explore available promotions

Description

Description

3-Methoxythiophenol is used in the preparation of allenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophenol is used in the preparation of allenes.

Solubility
Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.

Notes
Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

3-Methoxythiophenol
1.136
96°C (204°F)
C7H8OS
MFCD00004841
2041496
3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol
QMVAZEHZOPDGHA-UHFFFAOYSA-N
3-methoxybenzenethiol
84989
98%
15570-12-4
225°C to 227°C
Unpleasant
1.5865
5 g
Air Sensitive
Not miscible or difficult to mix in water. Soluble in methanol,benzene,hexane,toluene and dichloromethane.
COC1=CC(=CC=C1)S
140.2
140.21

Chemical Identifiers

15570-12-4
140.2
QMVAZEHZOPDGHA-UHFFFAOYSA-N
84989
COC1=CC(=CC=C1)S
C7H8OS
MFCD00004841
3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol
3-methoxybenzenethiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 239-617-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only