missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophene-2-carboxaldehyde, 97%

Catalog No. p-7032469
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33940MD 250 mg
AAH3394003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33940MD Supplier Thermo Scientific Chemicals Supplier No. H33940MD
Only null left

CAS: 35134-07-7 | C6H6O2S | 142.17 g/mol

3-Methoxythiophene-2-carboxaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophene-2-carboxaldehyde is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents. Store under inert gas.

Chemical Identifiers

CAS 35134-07-7
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD01571098
InChI Key KGJDTMQUUPIAEF-UHFFFAOYSA-N
PubChem CID 588315
IUPAC Name 3-methoxythiophene-2-carbaldehyde
SMILES COC1=C(SC=C1)C=O

Specifications

Melting Point 80°C to 82°C
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 3-methoxy-2-thiophenecarboxaldehyde, 3-methoxythiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methoxy, 3-methoxy-2-thenaldehyde, 2-formyl-3-methoxythiophene, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thiophenecarbaldehyde #
Solubility Information Slightly soluble in water.
Formula Weight 142.18
Percent Purity 97%
Chemical Name or Material 3-Methoxythiophene-2-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only