accessibility menu, dialog, popup

UserName

H3COSO

CAS RN 35134-07-7

IUPAC Name: 3-methoxythiophene-2-carbaldehyde
Synonyms: 3-methoxythiophene-2-carbaldehyde
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: KGJDTMQUUPIAEF-UHFFFAOYSA-N
SMILES: COC1=C(SC=C1)C=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)