missing translation for 'onlineSavingsMsg'
Learn More

3-methoxythiophene-2-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 35134-07-7
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD01571098
InChI Key KGJDTMQUUPIAEF-UHFFFAOYSA-N
Synonym 3-methoxy-2-thiophenecarboxaldehyde, 3-methoxythiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methoxy, 3-methoxy-2-thenaldehyde, 2-formyl-3-methoxythiophene, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thiophenecarbaldehyde #
PubChem CID 588315
IUPAC Name 3-methoxythiophene-2-carbaldehyde
SMILES COC1=C(SC=C1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC68604CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC68604DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

35134-07-7
142.17
KGJDTMQUUPIAEF-UHFFFAOYSA-N
588315
COC1=C(SC=C1)C=O
C6H6O2S
MFCD01571098
3-methoxy-2-thiophenecarboxaldehyde, 3-methoxythiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methoxy, 3-methoxy-2-thenaldehyde, 2-formyl-3-methoxythiophene, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thiophenecarbaldehyde #
3-methoxythiophene-2-carbaldehyde

Specifications

35134-07-7
100.0
C6H6O2S
3-methoxy-2-thiophenecarboxaldehyde, 3-methoxythiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methoxy, 3-methoxy-2-thenaldehyde, 2-formyl-3-methoxythiophene, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thiophenecarbaldehyde #
COC1=C(SC=C1)C=O
142.17
142.18
3-Methoxythiophene-2-carbaldehyde
96.5
97%
MFCD01571098
KGJDTMQUUPIAEF-UHFFFAOYSA-N
3-methoxythiophene-2-carbaldehyde
588315
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents