missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythioanisole 98.0+%, TCI America™

Catalog No. M18805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18805G 5 g
M188025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18805G Supplier TCI America Supplier No. M18805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2388-74-1
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD03789162
InChI Key IWVQVOXDIOKVBE-UHFFFAOYSA-N
Synonym 3-(Methylthio)anisole
PubChem CID 281182
IUPAC Name 1-methoxy-3-(methylsulfanyl)benzene
SMILES COC1=CC=CC(SC)=C1

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxythioanisole