missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxysalicylic Acid 98.0+%, TCI America™

Catalog No. p-7132565
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11535G 5 g
M115325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11535G Supplier TCI America Supplier No. M11535G
Only null left

Chemical Identifiers

CAS 877-22-5
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002445
InChI Key AUZQQIPZESHNMG-UHFFFAOYSA-N
Synonym 3-methoxysalicylic acid, o-vanillic acid, acide orthovanillique, benzoic acid, 2-hydroxy-3-methoxy, m-anisic acid, 2-hydroxy, 3-hydroxy-m-anisic acid, unii-3j31y0e2im, 2-hydroxy-3-methoxy-benzoic acid, 3-methoxysalicyclic acid, 2-hydroxy-3-methoxy benzoic acid
PubChem CID 70140
ChEBI CHEBI:68496
IUPAC Name 2-hydroxy-3-methoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1O

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxysalicylic Acid
RTECSNumber BZ4840000
TSCA No