missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxypyridazine 97.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 87°C |
| Quantity | 1 g |
| Formula Weight | 110.12 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Methoxypyridazine |
Safety and Handling
| TSCA | No |