accessibility menu, dialog, popup

UserName

H3CONN

CAS RN 19064-65-4

IUPAC Name: 3-methoxypyridazine
Synonyms: pyridazine, 3-methoxy-
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: ASFHDLDAWYTMJS-UHFFFAOYSA-N
SMILES: COC1=NN=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results