missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionic acid, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 453320500

Quantity 50mL

 View more versions of this product

Catalog No. AC453320500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2544-06-1
104.11
3-methoxypropionic acid, propanoic acid, 3-methoxy, propanoic acid, methoxy, m-peg1-acid, propanoic acid,methoxy, acmc-1aiwk, 3-methoxy-propionic acid, #, propanoic acid,3-methoxy, .beta.-methoxypropionic acid
3-methoxypropanoic acid
C4H8O3
YSIKHBWUBSFBRZ-UHFFFAOYSA-N
134442
COCCC(=O)O

Specifications

3-Methoxypropionic acid
1.1080g/mL
113°C
97.5% min. (GC)
50mL
3-methoxypropionic acid, propanoic acid, 3-methoxy, propanoic acid, methoxy, m-peg1-acid, propanoic acid,methoxy, acmc-1aiwk, 3-methoxy-propionic acid, #, propanoic acid,3-methoxy, .beta.-methoxypropionic acid
COCCC(=O)O
104.11
104.11
2544-06-1
116.0°C (9.0 mmHg)
Authentic
C4H8O3
1.108
YSIKHBWUBSFBRZ-UHFFFAOYSA-N
3-methoxypropanoic acid
134442
97+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...