accessibility menu, dialog, popup

UserName

CAS RN 2544-06-1

IUPAC Name: 3-methoxypropanoic acid
Synonyms: 3-methoxypropanoic acid
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: YSIKHBWUBSFBRZ-UHFFFAOYSA-N
SMILES: COCCC(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)