missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionic acid, 97+%, Thermo Scientific™

Catalog No. AC453320100
Change view
Click to view available options
Quantity:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453320100 10mL
AC453320500 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453320100 Supplier Thermo Scientific Chemicals Supplier No. 453320100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2544-06-1
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
InChI Key YSIKHBWUBSFBRZ-UHFFFAOYSA-N
Synonym 3-methoxypropionic acid, propanoic acid, 3-methoxy, propanoic acid, methoxy, m-peg1-acid, propanoic acid,methoxy, acmc-1aiwk, 3-methoxy-propionic acid, #, propanoic acid,3-methoxy, .beta.-methoxypropionic acid
PubChem CID 134442
IUPAC Name 3-methoxypropanoic acid
SMILES COCCC(=O)O

Specifications

Density 1.1080g/mL
Boiling Point 116.0°C (9.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 10mL
Specific Gravity 1.108
Formula Weight 104.11
Percent Purity 97+%
Chemical Name or Material 3-Methoxypropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only