missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylglyoxal hydrate, 97%, dry wt. basis, Thermo Scientific Chemicals

Catalog No. p-7048124
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2026303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2026303 Supplier Thermo Scientific Chemicals Supplier No. L2026303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32025-65-3 | Molecular Formula: C9H8O3 | Molecular Weight: 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32025-65-3
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD08533286
InChI Key SIHYQEYAJMDKQH-UHFFFAOYSA-N
Synonym 3-methoxyphenylglyoxal hydrate, 2-3-methoxyphenyl-2-oxoacetaldehyde hydrate, acmc-1c24s, bestipharma 555-843, 3-methoxyphenylglyoxal monohydrate, ethanone,2,2-dihydroxy-1-3-methylphenyl, 3-methoxyphenyl oxo acetaldehyde-water 1/1, 2-3-methoxyphenyl-2-oxidanylidene-ethanal hydrate
PubChem CID 21155397
IUPAC Name 2-(3-methoxyphenyl)-2-oxoacetaldehyde;hydrate
SMILES COC1=CC=CC(=C1)C(=O)C=O

Specifications

Melting Point 94°C to 98°C
Boiling Point 95°C to 98°C (0.05 mmHg)
Quantity 1 g
Beilstein 1935225
Formula Weight 164.16 (Anhydrous)
Percent Purity 97%
Assay (dry wt.)
Chemical Name or Material 3-Methoxyphenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only