missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxyphenylacetonitrile 98.0+%, TCI America™
$61.51 - $197.59
Chemical Identifiers
| CAS | 19924-43-7 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.177 |
| MDL Number | MFCD00001910 |
| InChI Key | LXKNAUOWEJWGTE-UHFFFAOYSA-N |
| Synonym | 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile |
| PubChem CID | 88310 |
| IUPAC Name | 2-(3-methoxyphenyl)acetonitrile |
| SMILES | COC1=CC=CC(=C1)CC#N |
Chemical Identifiers
| 19924-43-7 | |
| 147.177 | |
| LXKNAUOWEJWGTE-UHFFFAOYSA-N | |
| 88310 | |
| COC1=CC=CC(=C1)CC#N |
| C9H9NO | |
| MFCD00001910 | |
| 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile | |
| 2-(3-methoxyphenyl)acetonitrile |
Specifications
| 19924-43-7 | |
| 131°C | |
| MFCD00001910 | |
| 3276 | |
| LXKNAUOWEJWGTE-UHFFFAOYSA-N | |
| 2-(3-methoxyphenyl)acetonitrile | |
| 88310 | |
| ≥98.0% (GC) | |
| 3-Methoxyphenylacetonitrile |
| Yellow | |
| C9H9NO | |
| 5 g | |
| 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile | |
| COC1=CC=CC(=C1)CC#N | |
| 147.177 | |
| 147.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-1786
TSCA : No