missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetonitrile 98.0+%, TCI America™

Catalog No. M23315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23315G 5 g
M233125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23315G Supplier TCI America Supplier No. M23315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19924-43-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001910
InChI Key LXKNAUOWEJWGTE-UHFFFAOYSA-N
Synonym 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile
PubChem CID 88310
IUPAC Name 2-(3-methoxyphenyl)acetonitrile
SMILES COC1=CC=CC(=C1)CC#N

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 g
UN Number 3276
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenylacetonitrile
EINECSNumber (3)-1786
TSCA No