Learn More
3-Methoxyphenylacetonitrile, 99%
CAS: 19924-43-7 | C9H9NO | 147.177 g/mol
$79.08 - $692.00
Chemical Identifiers
| CAS | 19924-43-7 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.177 |
| MDL Number | MFCD00001910 |
| InChI Key | LXKNAUOWEJWGTE-UHFFFAOYSA-N |
| Synonym | 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile |
| PubChem CID | 88310 |
| IUPAC Name | 2-(3-methoxyphenyl)acetonitrile |
| SMILES | COC1=CC=CC(=C1)CC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1431706
|
Thermo Scientific Chemicals
A1431706 |
5 g |
Each for $79.08
|
|
|||||
|
AAA1431714
|
Thermo Scientific Chemicals
A1431714 |
25 g |
Each for $229.49
|
|
|||||
|
AAA1431722
|
Thermo Scientific Chemicals
A1431722 |
100 g |
Each for $692.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19924-43-7 | |
| 147.177 | |
| LXKNAUOWEJWGTE-UHFFFAOYSA-N | |
| 88310 | |
| COC1=CC=CC(=C1)CC#N |
| C9H9NO | |
| MFCD00001910 | |
| 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile | |
| 2-(3-methoxyphenyl)acetonitrile |
Specifications
| 19924-43-7 | |
| 278°C to 282°C | |
| C9H9NO | |
| MFCD00001910 | |
| UN3276 | |
| 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile | |
| COC1=CC=CC(=C1)CC#N | |
| 147.177 | |
| 147.18 | |
| 3-Methoxyphenylacetonitrile |
| 1.082 | |
| 98°C (208°F) | |
| 1.5312 | |
| 5 g | |
| 1865539 | |
| LXKNAUOWEJWGTE-UHFFFAOYSA-N | |
| 2-(3-methoxyphenyl)acetonitrile | |
| 88310 | |
| 99% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 243-428-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only