missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetonitrile, 99%

Catalog No. AAA1431706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1431706 5 g
AAA1431714 25 g
AAA1431722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1431706 Supplier Thermo Scientific Chemicals Supplier No. A1431706
Only null left

CAS: 19924-43-7 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19924-43-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001910
InChI Key LXKNAUOWEJWGTE-UHFFFAOYSA-N
Synonym 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile
PubChem CID 88310
IUPAC Name 2-(3-methoxyphenyl)acetonitrile
SMILES COC1=CC=CC(=C1)CC#N

Specifications

Density 1.082
Boiling Point 278°C to 282°C
Flash Point 98°C (208°F)
Refractive Index 1.5312
Quantity 5 g
UN Number UN3276
Beilstein 1865539
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 3-Methoxyphenylacetonitrile
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 243-428-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only