missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetone, 97%

Catalog No. AAB2091606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2091606 5 g
AAB2091603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2091606 Supplier Thermo Scientific Chemicals Supplier No. B2091606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3027-13-2 | C10H12O2 | 164.20 g/mol

3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 3027-13-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008771
InChI Key RMMRRRLPDBJBQL-UHFFFAOYSA-N
Synonym 3-methoxyphenylacetone, 1-3-methoxyphenyl propan-2-one, 1-3-methoxyphenyl acetone, 3-methoxyphenyl acetone, m-methoxyphenyl acetone, m-methoxyphenylacetone, 2-propanone, 1-3-methoxyphenyl, 3-methoxyphenyl methyl ketone, acmc-1ai0k
PubChem CID 76410
IUPAC Name 1-(3-methoxyphenyl)propan-2-one
SMILES COC1=CC=CC(CC(C)=O)=C1

Specifications

Density 1.078
Boiling Point 258°C to 260°C
Flash Point >110°C (230°F)
Refractive Index 1.5252
Quantity 5 g
Beilstein 2044357
Solubility Information Miscible with dimethyl sulfoxide.
Formula Weight 164.2
Percent Purity 97%
Chemical Name or Material 3-Methoxyphenylacetone
EINECSNumber 221-191-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only