missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. M085825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M085825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M085825G Supplier TCI America Supplier No. M085825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1798-09-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00004334
InChI Key LEGPZHPSIPPYIO-UHFFFAOYSA-N
Synonym 3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my
PubChem CID 15719
IUPAC Name 2-(3-methoxyphenyl)acetic acid
SMILES COC1=CC=CC(=C1)CC(=O)O

Specifications

Melting Point 69°C
Boiling Point 306°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxyphenylacetic Acid
RTECSNumber AI8940000
TSCA No