missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenyl isothiocyanate, 98%

Catalog No. AAA1509006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1509006 5 g
AAA1509014 25 g
AAA1509022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1509006 Supplier Thermo Scientific Chemicals Supplier No. A1509006

CAS: 3125-64-2 | C8H7NOS | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3125-64-2
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00040335
InChI Key WHBYCPUKGYEYFU-UHFFFAOYSA-N
Synonym 3-methoxyphenyl isothiocyanate, 3-methoxyphenylisothiocyanate, 1-isothiocyanato-3-methoxy-benzene, 3-isothiocyanatoanisole, acmc-1cldj, m-methoxyphenyl isothiocyanate, 3-methoxy phenyl isothiocyanate, 3-methoxy-phenyl isothiocyanate, #
PubChem CID 137832
IUPAC Name 1-isothiocyanato-3-methoxybenzene
SMILES COC1=CC=CC(=C1)N=C=S

Specifications

Density 1.179
Boiling Point 133°C to 134°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.639
Quantity 5 g
UN Number UN2922
Beilstein 775987
Sensitivity Moisture sensitive
Formula Weight 165.22
Percent Purity 98%
Chemical Name or Material 3-Methoxyphenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only