missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenyl Isocyanate 97.0+%, TCI America™

Catalog No. p-7131478
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05515G 5 g
I055125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05515G Supplier TCI America Supplier No. I05515G
Only null left

Chemical Identifiers

CAS 18908-07-1
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00002019
InChI Key NPOVTGVGOBJZPY-UHFFFAOYSA-N
Synonym 3-methoxyphenyl isocyanate, benzene, 1-isocyanato-3-methoxy, isocyanic acid 3-methoxyphenyl ester, 3-isocyanatoanisole, 3-methoxyphenylisocyanate, m-methoxyphenyl isocyanate, 1-isocyanato-3-methoxy-benzene, 3-methoxybenzenisocyanate, m-anisyl isocyanate, m-methoxyphenylisocyanate
PubChem CID 87843
IUPAC Name 1-isocyanato-3-methoxybenzene
SMILES COC1=CC=CC(=C1)N=C=O

Specifications

Color Colorless
Boiling Point 95°C
Quantity 5 g
UN Number 2206
Formula Weight 149.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator