missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenyl isocyanate, 99%

Catalog No. AC221160250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221160250 25 g Glass bottle
AC221160050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221160250 Supplier Thermo Scientific Chemicals Supplier No. 221160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18908-07-1 | C8H7NO2 | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18908-07-1
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00002019
InChI Key NPOVTGVGOBJZPY-UHFFFAOYSA-N
Synonym 3-methoxyphenyl isocyanate, benzene, 1-isocyanato-3-methoxy, isocyanic acid 3-methoxyphenyl ester, 3-isocyanatoanisole, 3-methoxyphenylisocyanate, m-methoxyphenyl isocyanate, 1-isocyanato-3-methoxy-benzene, 3-methoxybenzenisocyanate, m-anisyl isocyanate, m-methoxyphenylisocyanate
PubChem CID 87843
IUPAC Name 1-isocyanato-3-methoxybenzene
SMILES COC1=CC=CC(=C1)N=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Pink
Density 1.1300g/mL
Boiling Point 94°C to 95°C (10.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5420 to 1.544
Linear Formula CH3OC6H4NCO
Quantity 25 g
Beilstein 13, III, 963
Specific Gravity 1.13
Formula Weight 149.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenyl isocyanate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 242-658-3
TSCA TSCA

RUO – Research Use Only