missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenyl isocyanate, 99%

Catalog No. AAB2432906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2432906 5 g
AAB2432914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2432906 Supplier Thermo Scientific Chemicals Supplier No. B2432906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18908-07-1 | C8H7NO2 | 149.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18908-07-1
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00002019
InChI Key NPOVTGVGOBJZPY-UHFFFAOYSA-N
Synonym 3-methoxyphenyl isocyanate, benzene, 1-isocyanato-3-methoxy, isocyanic acid 3-methoxyphenyl ester, 3-isocyanatoanisole, 3-methoxyphenylisocyanate, m-methoxyphenyl isocyanate, 1-isocyanato-3-methoxy-benzene, 3-methoxybenzenisocyanate, m-anisyl isocyanate, m-methoxyphenylisocyanate
PubChem CID 87843
IUPAC Name 1-isocyanato-3-methoxybenzene
SMILES COC1=CC=CC(=C1)N=C=O

Specifications

Density 1.138
Boiling Point 94°C to 95°C (10 mmHg)
Flash Point 96°C (204°F)
Odor Pungent
Refractive Index 1.543
Quantity 5 g
UN Number UN2206
Beilstein 775986
Sensitivity Moisture sensitive
Formula Weight 149.15
Percent Purity 99%
Chemical Name or Material 3-Methoxyphenyl isocyanate
Show More Show Less
Hazard Category H302-H312-H315-H319-H332-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 242-658-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only