missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenoxyacetic acid, 97+%

Catalog No. p-7043516
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0110006 5 g
AAL0110014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0110006 Supplier Thermo Scientific Chemicals Supplier No. L0110006

CAS: 2088-24-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2088-24-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00014357
InChI Key AHDPQRIYMMZJTF-UHFFFAOYSA-N
Synonym 3-methoxyphenoxy acetic acid, 3-methoxyphenoxyacetic acid, 2-3-methoxyphenoxy acetic acid, acetic acid, 3-methoxyphenoxy, m-methoxy phenoxy acetic acid, m-methoxyphenoxyacetic acid, aurora 22719, acmc-20an1j, 3-methoxyphenoxy-acetic acid, 3-methoxy-phenoxy-acetic acid
PubChem CID 74969
IUPAC Name 2-(3-methoxyphenoxy)acetic acid
SMILES COC1=CC=CC(OCC(O)=O)=C1

Specifications

Melting Point 116°C to 119°C
Quantity 5 g
Beilstein 1876898
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxyphenoxyacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only