missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenol, 97%

Catalog No. aaa1282614
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAA1282614 25 g
AAA1282622 100 g
AAA1282636 500 g
3 options

Catalog No. AAA1282614

Supplier: Thermo Scientific Chemicals A1282614

Only null left

CAS: 150-19-6 | C7H8O2 | 124.139 g/mol

It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

Solubility
Partly soluble in water.

Notes
Store away from oxidizing agents, acids, bases, acid chlorides and acid anhydrides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources

Chemical Identifiers

CAS 150-19-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002267
InChI Key ASHGTJPOSUFTGB-UHFFFAOYSA-N
Synonym m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
PubChem CID 9007
ChEBI CHEBI:52678
IUPAC Name 3-methoxyphenol
SMILES COC1=CC=CC(=C1)O

Specifications

Melting Point -18°C
Density 1.14
Boiling Point 244°C
Flash Point 124°C (255°F)
Odor Sweet
Refractive Index 1.552
Quantity 25 g
UN Number UN2810
Beilstein 1209898
Solubility Information Partly soluble in water.
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 3-Methoxyphenol
Show More Show Less
Hazard Category H302+H332-H311-H315-H318-H335
Hazard Statement GHS H Statement
H311-H318-H302-H332-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 205-754-6
RTECSNumber SL7524000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only