missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenol, 97%

Catalog No. AC161060250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161060250 25 mL Glass bottle
AC161061000 100 mL Glass bottle
AC161065000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161060250 Supplier Thermo Scientific Chemicals Supplier No. 161060250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 150-19-6 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-19-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002267
InChI Key ASHGTJPOSUFTGB-UHFFFAOYSA-N
Synonym m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
PubChem CID 9007
ChEBI CHEBI:52678
IUPAC Name 3-methoxyphenol
SMILES COC1=CC=CC(=C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -17.0°C
Color Brown to Pink-Red
Density 1.1450g/mL
Boiling Point 244.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5510 to 1.5530
Linear Formula CH3OC6H4OH
Quantity 25 mL
Beilstein 06, 813
Fieser 13,181
Specific Gravity 1.145
Solubility Information Solubility in water: slightly soluble
Formula Weight 124.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doc
EINECSNumber 205-754-6

RUO – Research Use Only