missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenethylamine, 98+%

Catalog No. AC302610250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302610250 25 g Glass bottle
AC302610050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC302610250 Supplier Thermo Scientific Chemicals Supplier No. 302610250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2039-67-0 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2039-67-0
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00008187
InChI Key WJBMRZAHTUFBGE-UHFFFAOYSA-N
Synonym 3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
PubChem CID 74866
IUPAC Name 2-(3-methoxyphenyl)ethanamine
SMILES COC1=CC=CC(=C1)CCN

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0380g/mL
Boiling Point 118°C to 119°C (6.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5375 to 1.5395 (20°C, 589nm)
Quantity 25 g
Beilstein 13, IV, 1781
Specific Gravity 1.038
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 151.21
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 218-017-9

RUO – Research Use Only