missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxydiphenylamine 98.0+%, TCI America™

Catalog No. M1135500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1135500G 500 g
M113525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1135500G Supplier TCI America Supplier No. M1135500G

Chemical Identifiers

CAS 101-16-6
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00008383
InChI Key MKASXAGBWHIGCF-UHFFFAOYSA-N
Synonym 3-methoxydiphenylamine, n-phenyl-m-anisidine, benzenamine, 3-methoxy-n-phenyl, n-phenyl-3-methoxyaniline, n1-phenyl-3-methoxyaniline, n-3-methoxyphenyl-n-phenylamine, 3-methoxy-n-phenylbenzenamine, 3-methoxydiphenylamin, 3-methoxy diphenylamine, pubchem13530
PubChem CID 7544
IUPAC Name 3-methoxy-N-phenylaniline
SMILES COC1=CC=CC(=C1)NC2=CC=CC=C2

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 160°C
Quantity 500 g
Formula Weight 199.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxydiphenylamine
EINECSNumber (3)-2788
TSCA No