missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxycinnamic Acid 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 119°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 178.19 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Methoxycinnamic Acid |
Safety and Handling
| TSCA | No |