missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxycatechol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 42°C |
| Color | White-Yellow |
| Boiling Point | 164°C |
| Quantity | 25 g |
| Formula Weight | 140.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Methoxycatechol |
Safety and Handling
| RTECSNumber | CZ9002750 |
| TSCA | No |
| Recommended Storage | Refrigerator |