Learn More
3-(Methoxycarbonyl)phenyl isocyanate, 97%
CAS: 41221-47-0 | C9H7NO3 | 177.16 g/mol
$120.21 - $411.32
Chemical Identifiers
| CAS | 41221-47-0 |
|---|---|
| Molecular Formula | C9H7NO3 |
| Molecular Weight (g/mol) | 177.16 |
| MDL Number | MFCD00037077 |
| InChI Key | WBGWGERFPSYHDT-UHFFFAOYSA-N |
| Synonym | 3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate |
| PubChem CID | 2733366 |
| IUPAC Name | methyl 3-isocyanatobenzoate |
| SMILES | COC(=O)C1=CC=CC(=C1)N=C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1870903
|
Thermo Scientific Chemicals
L1870903 |
1 g |
Each for $120.21
|
|
|||||
|
AAL1870906
|
Thermo Scientific Chemicals
L1870906 |
5 g |
Each for $411.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 41221-47-0 | |
| 177.16 | |
| WBGWGERFPSYHDT-UHFFFAOYSA-N | |
| 2733366 | |
| COC(=O)C1=CC=CC(=C1)N=C=O |
| C9H7NO3 | |
| MFCD00037077 | |
| 3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate | |
| methyl 3-isocyanatobenzoate |
Specifications
| 41221-47-0 | |
| >110°C (230°F) | |
| MFCD00037077 | |
| UN2811 | |
| 3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate | |
| COC(=O)C1=CC=CC(=C1)N=C=O | |
| 177.16 | |
| 177.16 | |
| 3-(Methoxycarbonyl)phenyl isocyanate |
| 35°C to 38°C | |
| C9H7NO3 | |
| 1 g | |
| Moisture sensitive | |
| WBGWGERFPSYHDT-UHFFFAOYSA-N | |
| methyl 3-isocyanatobenzoate | |
| 2733366 | |
| 97% |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
H302+H312+H332-H315-H317-H319-H334-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only