CAS RN 41221-47-0
IUPAC Name:
methyl 3-isocyanatobenzoate
Synonyms:
methyl 3-isocyanatobenzoate
Molecular Weight (g/mol):
177.16
Molecular Formula:
C9H7NO3
InChi Key:
WBGWGERFPSYHDT-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC(=C1)N=C=O
1
–
1
of
1
results
1
Promotions Available
3-(Methoxycarbonyl)phenyl isocyanate, 97%
3-(Methoxycarbonyl)phenyl isocyanate, 97%
CAS: 41221-47-0 Molecular Formula: C9H7NO3 Molecular Weight (g/mol): 177.16 MDL Number: MFCD00037077 InChI Key: WBGWGERFPSYHDT-UHFFFAOYSA-N Synonym: 3-carbomethoxyphenyl isocyanate,3-isocyanatobenzoic acid methyl ester,benzoic acid, 3-isocyanato-, methyl ester,3-methoxycarbonyl phenyl isocyanate,acmc-20akh8,methyl-3-isocyanatobenzoate,3-carbomethoxyphenylisocyanate,methyl 3-isocyanatobenzoate,3-isocyanato-benzoic acid methyl ester,m-methoxycarbonylphenyl isocyanate PubChem CID: 2733366 IUPAC Name: methyl 3-isocyanatobenzoate SMILES: COC(=O)C1=CC=CC(=C1)N=C=O