missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybutyl Thioglycolate 97.0+%, TCI America™

Catalog No. p-7132995
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
M206025G 25 g
M2060500G 500 g
2 options

Catalog No. M206025G

Supplier: TCI America M206025G

Only null left

Chemical Identifiers

CAS 27431-39-6
Molecular Formula C7H14O3S
Molecular Weight (g/mol) 178.246
MDL Number MFCD04038084
InChI Key FVOSQWCSFOEXOP-UHFFFAOYSA-N
Synonym Thioglycolic Acid 3-Methoxybutyl Ester, 3-Methoxybutyl Mercaptoacetate, Mercaptoacetic Acid 3-Methoxybutyl Ester
PubChem CID 11998744
IUPAC Name 3-methoxybutyl 2-sulfanylacetate
SMILES CC(CCOC(=O)CS)OC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 178.25
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methoxybutyl Thioglycolate
EINECSNumber (2)-1357
TSCA No