accessibility menu, dialog, popup

UserName

CAS RN 27431-39-6

CH3OOHSOCH3

CAS RN 27431-39-6

IUPAC Name: 3-methoxybutyl 2-sulfanylacetate
Synonyms: 3-methoxybutyl 2-sulfanylacetate
Molecular Weight (g/mol): 178.25
Molecular Formula: C7H14O3S
InChi Key: FVOSQWCSFOEXOP-UHFFFAOYNA-N
SMILES: COC(C)CCOC(=O)CS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results