missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxybutyl Thioglycolate 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 178.25 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Methoxybutyl Thioglycolate |
Safety and Handling
| EINECSNumber | (2)-1357 |
| TSCA | No |